Learn More
Kimble™ Kim-Kap™ Disposable Closures Available on GSA/VA Contract for Federal Government customers only.
Available in natural (clear) or choice of four colors
Manufacturer:  Kimble™ 7366416
 View more versions of this product
Catalog No. 14-957-92F
  Check Availability
Add to cart
Specifications
Description
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
Caps for 38mm O.D. are designed for baffled culture flasks.
Description & Specifications
Specifications
Yes
Polypropylene
Closure
16mm O.D. plain-end culture tubes
Yellow
Yellow
Yes