Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™-35 300mL BOD Bottles
SDP

For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis

Supplier:  DWK Life Sciences 1507025

 View more versions of this product

Catalog No. 02-885C


Description

Description

For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis

Flared mouth forms a water seal to prevent air from being drawn into the bottle during incubation, while shoulder radius provides an interior shape that sweeps entrapped air out of the stopper opening.

  • Type 1, Class B borosilicate glass
  • Available numbered and unnumbered With glass stopper unless otherwise noted
  • Optional snap lid secures stopper in bottle and reduces evaporation of water seal during incubation (sold separately, 02-939)
  • Size: 2.75 dia. × 6.5 in. H (69 × 165mm); neck I.D. approximately 0.66 in.  (16.7mm)

Order Info

Plastic snap lids not included. For these, see Cat. No. 02-939.

Compliance

Conforms to ASTM™ E 438

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ASTM E438 type I, class A
For sampling and incubation of aqueous samples
25 to 48
10.14 oz.
16.5 cm
6.9 cm
300 mL
Borosilicate Glass
With stopper
6.5 in.
2.7 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.