Learn More
DWK Life Sciences Kimble™ KIMAX™-35 300mL BOD Bottles
For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis
Supplier: DWK Life Sciences 1507000
Description
For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis
Flared mouth forms a water seal to prevent air from being drawn into the bottle during incubation, while shoulder radius provides an interior shape that sweeps entrapped air out of the stopper opening.
- Type 1, Class B borosilicate glass
- Available numbered and unnumbered With glass stopper unless otherwise noted
- Optional snap lid secures stopper in bottle and reduces evaporation of water seal during incubation (sold separately, 02-939)
- Size: 2.75 dia. × 6.5 in. H (69 × 165mm); neck I.D. approximately 0.66 in. (16.7mm)
Order Info
Plastic snap lids not included. For these, see Cat. No. 02-939.
Compliance
Conforms to ASTM™ E 438
Specifications
ASTM E438 type I, class A | |
For sampling and incubation of aqueous samples | |
Unnumbered | |
10.14 oz. | |
16.5 cm | |
6.9 cm |
300 mL | |
Borosilicate Glass | |
With stopper | |
6.5 in. | |
2.7 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.