Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ KIMAX™-51 Brand Reusable Class A Mohr (Measuring) Pipets Available on GSA/VA Contract for Federal Government customers only.

Subdivisions on all sizes are 0.1mL, except 13-664-10H where subdivisions are 0.01mL

Manufacturer:  Kimble™ 3702510

 View more versions of this product

Catalog No. 13-664-10L



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Measuring Pipets
0.33 oz., 10mL
A
Orange
0.1mL
350mm
±0.03mL
10mL
To deliver
ASTM E1293
Orange
No
13.7 in.
0.1mL
0.33 oz.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit