Learn More
Kimble™ KIMAX™ Automatic Burets with Bottles Available on GSA/VA Contract for Federal Government customers only.
Class A
Manufacturer:  Kimble™ 17124F10
 View more versions of this product
Catalog No. 03-848-2A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Automatic Buret
±0.02mL
Volumetric analysis
Reservoir bottle, U-shaped drying tube, vented connecting tube, rubber squeeze bulb, No. 1 single-holed rubber stopper, PTFE stopcock plug and 1/4 in. I.D. rubber tubing, durable black ceramic enamel scale
765mm
29
0.05mL
NIST, ASTM E694, ASTM E542
0.5 to 10mL
30.1 in.
Borosilicate Glass
10mL