Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Class A Cylinders: PROMO
White scale; w/reverse graduations and bumper
Manufacturer:  DWK Life Sciences 20028W50PM
 View more versions of this product
Catalog No. 08-561-APM
Add to cart
Description
Description
  • Calibrated “To Deliver”
  • Double scale with reverse graduations
Glass Class A cylinders are calibrated “To Deliver” for greatest accuracy. Double metric scale with reverse graduations makes reading easy. White scales for darker samples. Hexagonal ground base provides stability.
Specifications
Specifications
Graduated 3 to 50mL
Borosilicate Glass
A
7.48 in.
±0.25mL
1mL
190mm
ASTM E438
50mL
Double white metric scale with reverse graduations
1mL