Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Class A Cylinders: PROMO
White scale; w/reverse graduations and bumper
Manufacturer:  DWK Life Sciences 20028W1000PM
 View more versions of this product
Catalog No. 08-568-APM
Add to cart
Description
Description
  • Calibrated “To Deliver”
  • Double scale with reverse graduations
Glass Class A cylinders are calibrated “To Deliver” for greatest accuracy. Double metric scale with reverse graduations makes reading easy. White scales for darker samples. Hexagonal ground base provides stability.
Specifications
Specifications
Graduated 50 to 1000mL
Borosilicate Glass
A
18.11 in.
±2.5mL
10mL
460mm
ASTM E438
1000mL
Double white metric scale with reverse graduations
10mL