Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Class A Cylinders: PROMO
White scale; w/reverse graduations and bumper
Manufacturer:  DWK Life Sciences 20028W500PM
 View more versions of this product
Catalog No. 08-567-APM
Add to cart
Description
Description
  • Calibrated “To Deliver”
  • Double scale with reverse graduations
Glass Class A cylinders are calibrated “To Deliver” for greatest accuracy. Double metric scale with reverse graduations makes reading easy. White scales for darker samples. Hexagonal ground base provides stability.
Specifications
Specifications
Graduated 25 to 500mL
Borosilicate Glass
A
14.76 in.
±1.3mL
5mL
375mm
ASTM E438
500mL
Double white metric scale with reverse graduations
5mL