Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Class A Cylinders: PROMO Available on GSA/VA Contract for Federal Government customers only.
White scale; w/reverse graduations and bumper
$120.00 - $337.00
Specifications
Material Borosilicate Glass
Certifications/Compliance ASTM E438
Class A
Scale Double white metric scale with reverse graduations
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity & Availability  
08-558-APM Available on GSA/VA Contract for Federal Government customers only. View Documents DWK Life Sciences
20028W10PM
10 mL 0.1 mL Case of 6 for $206.00
Add to cart
08-560-APM Available on GSA/VA Contract for Federal Government customers only. View Documents DWK Life Sciences
20028W25PM
25 mL 0.2 mL Case of 6 for $222.00
Add to cart
08-561-APM Available on GSA/VA Contract for Federal Government customers only. View Documents DWK Life Sciences
20028W50PM
50 mL 1 mL Case of 6 for $225.00
Add to cart
08-564-APM Available on GSA/VA Contract for Federal Government customers only. View Documents DWK Life Sciences
20028W250PM
250 mL 2 mL Case of 4 for $240.00
Add to cart
08-567-APM Available on GSA/VA Contract for Federal Government customers only. View Documents DWK Life Sciences
20028W500PM
500 mL 5 mL Case of 4 for $337.00
Add to cart
08-568-APM Available on GSA/VA Contract for Federal Government customers only. View Documents DWK Life Sciences
20028W1000PM
1000 mL 10 mL Case for $120.00
Add to cart
Description
Description
  • Calibrated “To Deliver”
  • Double scale with reverse graduations
Glass Class A cylinders are calibrated “To Deliver” for greatest accuracy. Double metric scale with reverse graduations makes reading easy. White scales for darker samples. Hexagonal ground base provides stability.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Borosilicate Glass
ASTM E438
A
Double white metric scale with reverse graduations