Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
DWK Life Sciences Kimble™ KIMAX™ Brand Class A Cylinders: PROMO
White scale; w/reverse graduations and bumper
Manufacturer:  DWK Life Sciences 20028W10PM
 View more versions of this product
Catalog No. 08-558-APM
Description
Description
  • Calibrated “To Deliver”
  • Double scale with reverse graduations
Glass Class A cylinders are calibrated “To Deliver” for greatest accuracy. Double metric scale with reverse graduations makes reading easy. White scales for darker samples. Hexagonal ground base provides stability.
Specifications
Specifications
Graduated 1 to 10mL
Borosilicate Glass
A
6.49 in.
±0.09mL
0.1mL
165mm
ASTM E438
10mL
Double white metric scale with reverse graduations
0.1mL