Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™ Colorware™ Low-Form Beakers, yellow

Kimax-33 glass beakers with brightly colored graduations and marking spot

Manufacturer:  DWK Life Sciences 14000Y150

 View more versions of this product

Catalog No. 02-539-18


Add to cart

Description

Description

  • With pouring spout
  • Matte-finish marking spot can be marked with ordinary pencil
  • Colors make graduated scale easy to read

Compliance

Designed from ASTM™ E 960 Type I requirements

Specifications

Specifications

5.07 oz.
Sunny Yellow
5.7cm
Yes
3.11 in.
20mL
12 Case
Beaker
Borosilicate Glass
ASTM, E960, Type I
2.24 in.
For easy identification in the lab, reduce cross contamination
20 to 140mL
7.9cm
Matte finish marking
Dual scale
150mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit