Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ KIMAX™ Colorware™ Low-Form Beakers, green

Kimax-33 glass beakers with brightly colored graduations and marking spot

Manufacturer:  Kimble™ 14000G600

 View more versions of this product

Catalog No. 10-200-21


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

12.2cm
600mL
Cool Green
3.46 in.
For easy identification in the lab, reduce cross contamination
50 to 500mL
Matte finish marking
Dual scale
20.28 oz.
4.8 in.
Borosilicate Glass
ASTM, E960, Type I
8.8cm
Yes
50mL
6 Case
20.28 oz., 600mL
Beaker
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit