Learn More
Kimble™ KIMAX™ Culture Tubes Products for Science Education
Manufacturer:  Kimble™ 450481075/EMD
 View more versions of this product
Catalog No. S30521
Add to cart
Description
Description
Durable Kimax glass tubes without lip, with fire-polished top rim!
Specifications
Specifications
USP Type I, ASTM E 438 Type I, Class A
White Ceramic Enamel Marking Spot
10 mm
Borosilicate Glass
No
4 mL
75 mm