Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ KIMAX™ Disposable Glass Tubes Available on GSA/VA Contract for Federal Government customers only.

Use these glass tubes for a variety of applications and cell washing procedures. Kimble™ KIMAX™ Disposable Glass Tubes are made from 51 expansion borosilicate glass, which offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products.

Manufacturer:  Kimble™ 7350020150

 View more versions of this product

Catalog No. 14-957-96K


  Check Availability
Add to cart

Specifications

Description

  • These premium quality tubes feature sturdy, uniform bottoms and consistent lengths 
  • The 10 x 75 and 12 x 75mm sizes are suitable for cell washing procedures
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • The 6 x 50mm size is often referred to as a “Durham tube”
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
  • No marking spot
  • Designed from ASTM Specification E890
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Description & Specifications

Specifications

USP Type I, ASTM E 438 Type I, Class B
Yes
None
36mL
150mm
Uniform
20mm
Excellent chemical resistance against sodium leaching
Tray
Without Cap
Borosilicate Glass
Promotions

Promotions

  • Kimble
    For a limited time, buy three cases of Kimble™ KIMAX™ Disposable or Reusable Glass Tubes and get a complimentary fourth case of tubes (equal or lesser value).

    06-30-2017

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit