Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™ Heavy-Duty Beakers

Heavy construction prolongs life

Manufacturer:  DWK Life Sciences 14005400

 View more versions of this product

Catalog No. 02-555-20B



  Check Availability
Add to cart

Alert:

Not recommended for use on hotplates with thermal gradients or without agitation.


Description

Description

Heavy construction prolongs life

  • With pouring spout
  • With marking spot
  • Have white graduations for approximating volume
  • Accurate to ±5%
  • Thick, uniform walls with extra wall thickness in tooled top rim

Specifications

Specifications

±5%
USP, ASTM E438, ASTM E960
7.7cm
Offer superior mechanical strength and durability, provides improved safety when used under extreme conditions such as mechanized washing and rough handling
25 to 325mL
11.4cm
25mL
12 Pack
Beaker
Borosilicate Glass
13.52 oz.
3.03 in.
No
Yes
4.48 in.
Spout
Matte finish marking
Dual white scale
400mL
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit