Learn More
Kimble™ KIMAX™ Heavy-duty Filtering Flask Products for Science Education
The plastic sidearm conveniently detaches from the flask
$77.00
Specifications
Material Borosilicate Glass
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S33662 Products for Science Education
View Documents
Kimble™
27065250/EMD
250 mL
Pack of 2 for $77.00
Case of 9 PK for $690.84
Add to cart
Description
Description
  • Made of heavy-duty Kimax borosilicate glass
  • Has a detachable and autoclavable plastic sidearm
  • Sidearm attached by neoprene grommet
Order Info
Sold in packs of 2
Specifications
Specifications
Borosilicate Glass