Learn More
DWK Life Sciences Kimble Chase™ KIMAX™ Lazer-Fused™ Oil Centrifuge Tube
Kimble-Chase KIMAX™ Lazer-Fused™ Oil Centrifuge Tube used in determination of water and sediment in crude mineral oils, fuel oils and other petroleum products.
Manufacturer:  DWK Life Sciences 45240LF100
Catalog No. 11-555-262
Add to cart
Description
Description
  • Calibrated to contain
  • Scale and legend are durable white ceramic enamel
  • Maximum RCF: 800