Learn More
DWK Life Sciences Kimble Chase™ KIMAX™ Lazer-Fused™ Short Cone Oil Centrifuge Tube
Designed for field use in testing petroleum
Manufacturer:  DWK Life Sciences 45243LF100
 View more versions of this product
Catalog No. 11-555-261
Add to cart
Description
Description
  • Calibrated to contain
  • Scale and legend are durable white ceramic enamel
  • Maximum RCF: 800