Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ KIMAX™ Microburets with PTFE-Plug Stopcock Available on GSA/VA Contract for Federal Government customers only.

Used for small volume titrations requiring traceable volumetric accuracy

Manufacturer:  Kimble™ 17110F5

 View more versions of this product

Catalog No. 20-105A


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.16 oz.
Used for small volume titrations requiring traceable volumetric accuracy
No. 2 PTFE-plug stopcock, white ceramic enamel scale
Borosilicate Glass
5mL
790mm
±0.01mL
USP Type I and ASTM E438, Type I, Class A, NIST
Yes
31.1 in.
PTFE
0.16 oz., 5mL
0.01mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit