Learn More
Kimble™ KIMAX™ Precision Bore Burets with PTFE Stopcock Available on GSA/VA Contract for Federal Government customers only.
Class A
Manufacturer:  Kimble™ 17027F10
 View more versions of this product
Catalog No. 03-700-22A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Buret
0.33 oz.
A
Case of 1
No. 2 stopcock and KimKap™ dust cap
66.4cm
White Ceramic Enamel
±0.02mL
0.33 oz., 10mL
0.05mL
ASTM E287, ASTM E694, ASTM E542, NIST, USP Type I, ASTM E438, Type I, Class A
For general purpose titrations requiring traceable volumetric accuracy
26.14 in.
Borosilicate Glass
PTFE
10mL