Learn More
DWK Life Sciences Kimble™ KIMAX™ Reagent Bottles Available on GSA/VA Contract for Federal Government customers only.
For accurate rubber stopper fit
Manufacturer:  DWK Life Sciences 150932000
 View more versions of this product
Catalog No. 02-886-8E
Add to cart
Description
Description
  • Borosilicate glass
  • Tooled top and narrow mouth
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Stoppers are available separately; order online
Compliance
Meet ASTM™ E 438-92, Type I, Class A requirements.
Specifications
Specifications
67.62 oz., 2000mL
67.62 oz.
Rubber
1.06 in.
Tooled neck
Borosilicate Glass
250mm
2000mL
ASTM E438-92, Type I, Class A
5.35 in.
27mm
9.84 in.
136mm
Narrow