Learn More
DWK Life Sciences Kimble™ KIMAX™ Reagent Bottles
For accurate rubber stopper fit
Manufacturer:  DWK Life Sciences 150935000
 View more versions of this product
Catalog No. 02-886-8F
Add to cart
Description
Description
  • Borosilicate glass
  • Tooled top and narrow mouth
Order Info
Stoppers are available separately; order online
Compliance
Meet ASTM™ E 438-92, Type I, Class A requirements.
Specifications
Specifications
169.07 oz., 5000mL
169.07 oz.
Rubber
1.65 in.
Tooled neck
Borosilicate Glass
320mm
5000 mL
ASTM E438-92, Type I, Class A
7.2 in.
42mm
12.58 in.
183mm
Narrow