Learn More
Kimble™ KIMAX™ Reusable Class A Volumetric Serialized Pipets
Autoclavable
Manufacturer:  Kimble™ 370102510
 View more versions of this product
Catalog No. 13-660P
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
2.5mL
Certificate of graduation accuracy, ASTM E969
±0.006mL
A
13.77 in.
Borosilicate Glass
No
To deliver
Serialized Volumetric Pipet
35cm