Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ KIMAX™ Reusable Class A Volumetric Serialized Pipets Available on GSA/VA Contract for Federal Government customers only.

Autoclavable

Manufacturer:  Kimble™ 370101

 View more versions of this product

Catalog No. 13-660A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Specifications

1mL
Certificate of graduation accuracy, ASTM E969
No
To deliver
Serialized Volumetric Pipet
31cm
Borosilicate Glass
Blue
±0.006mL
A
12.2 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit