Learn More
DWK Life Sciences Kimble™ Reusable Class A Volumetric Serialized and Certified Pipets, TD
SDP
Designed from ASTM Specification E969, Class A requirements. Kimble™ Reusable Class A Volumetric Serialized and Certified Pipets, TD are ideal for measuring accurate volumes of liquids.
Supplier:  DWK Life Sciences 370061510
 View more versions of this product
Catalog No. 02-555-346
Add to Cart
Description
Description
Designed from ASTM Specification E969, Class A requirements. Kimble™ KIMAX™ Reusable Class A Volumetric Serialized Pipets, TD are ideal for measuring accurate volumes of liquids.
  • Sizes 1mL and larger are marked with large numerals on the bulb, indicating capacity
  • Color-coded (ASTM E1273) for ease in sorting and selecting the correct pipet
  • With a legend of permanent brown stain
  • Permanently marked
Specifications
Specifications
Autoclavable
Certificate of graduation accuracy, ASTM E969
Borosilicate Glass
1.5 mL
Serialized Volumetric Pipet
To Deliver
A
±0.006 mL
33.5 cm
Reusable
Product Suggestions
Product Suggestions