Learn More
Kimble™ KIMAX™ Reusable Class A Volumetric Standard Pipets Available on GSA/VA Contract for Federal Government customers only.
Autoclavable
Manufacturer:  Kimble™ 3700420
 View more versions of this product
Catalog No. 13-660-5H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
20mL
ASTM E969
Yellow
±0.03mL
A
Glass
Yes
No
To deliver
Standard Volumetric Pipet