Learn More
DWK Life Sciences Kimble™ Kimble™ KIMAX™ Reusable Class A Volumetric Unserialized Pipets, TD
Calibrated to deliver. Kimble™ Reusable Class A Volumetric Unserialized Pipets, TD are designed to indicate the volume delivered from the vessel.
Manufacturer:  DWK Life Sciences 370022510
 View more versions of this product
Catalog No. 02-555-313
Add to cart
Description
Description
  • Color coded
  • Designed from ASTM Specification E969, Class A requirements
  • 75 and 200mL sizes are designed from ASTM E542
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class
Specifications
Specifications
Yes
0.084 oz., 2.5mL
A
35cm
Unserialized Volumetric Pipet
To Contain, To Deliver
ASTM E969
No
±0.006mL
Borosilicate Glass