Kimble™ KIMAX™ Reusable Tubes with PTFE-Faced Rubber-Lined Screw Cap Available on GSA/VA Contract for Federal Government customers only.

Black phenolic cap with PTFE-faced rubber liner

Manufacturer: Kimble™ 45066A25150

 View more versions of this product

Catalog No. 14-930-10J

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM E982, Type VI, Class B
    Closure Color Black
    Closure Material Phenolic
    Label Type Marking Spot
    Liner Material PTFE, Rubber
    Thread Size 24-410
    Capacity (Metric) 50mL
    Closure Type Screw
    Diameter (Metric) Outer 25mm
    Length (Metric) 150mm
    Material Borosilicate Glass
    Shape Round Bottom

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit