Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™ Reusable Volumetric Unserialized Class A Pipets: To Deliver

Kimax rugged construction volumetric pipet; calibrated to deliver.

$96.00

Specifications

Class A
Disposable No
Material Borosilicate Glass
Type Unserialized Volumetric Pipet
View More Specs

Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
13-660-6E
View Documents
DWK Life Sciences
37004200
200 mL
Each for $96.00
Case of 6 EA for $575.75
Add to cart
 
Description

Description

Color-coded (ASTM E1273) for ease in sorting and selecting the correct pipet.
Specifications

Specifications

A
No
Borosilicate Glass
Unserialized Volumetric Pipet
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit