Learn More
Kimble™ KIMAX™ Schellbach Burets with Three-Way Stopcock Available on GSA/VA Contract for Federal Government customers only.
With No. 2 three-way stopcock with automatic PTFE plug
Manufacturer:  Kimble™ 17170F25
 View more versions of this product
Catalog No. 03-750-10A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.84 oz.
1 to 25mL
23.38 in.
Borosilicate Glass
PTFE
25mL
Buret
ASTM E287, USP Type I and ASTM E438, Type I, Class A
No. 2 three-way stopcock, automatic PTFE-plug
59.4cm
Black Ceramic Enamel
±0.06mL
0.84 oz., 25mL
0.1mL