Learn More
Kimble™ KIMAX™ Test Tubes with Lip and Marking Spot
Durable Kimax borosilicate tubes.
Supplier: Kimble™ 4504218150/EMD
This item is not returnable.
View return policy
Description
Quality glass test tubes with lip and ceramic enamel marking spot.
Specifications
ASTM E982, Type I | |
28 mL | |
Borosilicate Glass | |
18 mm |
Plain | |
150 mm | |
Marking Spot |