Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ KIMAX™/ValueWare™ 58° Angle Short Stem Funnels
SCI_ED

58° angle provides faster, more effective filtering.

Manufacturer:  Kimble™ 2895075/EMD

 View more versions of this product

Catalog No. S34404



Add to cart

Description

Description

Specifications

Specifications

7.5cm
0.8cm
ASTM E1095, Type I, Class A
12.5cm
7.5cm
90 mL
Borosilicate Glass
58°
Provides faster, more effective filtering
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit