Learn More
Kimble KIMAX/ValueWare 58° Angle Short Stem Funnels
58° angle provides faster, more effective filtering.
$86.00 - $248.00
Specifications
Angle | 58° |
---|---|
Certifications/Compliance | ASTM E1095, Type I, Class A |
Material | Borosilicate Glass |
For Use With (Application) | Provides faster, more effective filtering |
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Stem | Diameter (Metric)Top | Length (Metric) Stem | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Stem | Diameter (Metric)Top | Length (Metric) Stem | Price | Quantity & Availability | ||
S34401
![]() |
Kimble
2895045/EMD |
19 mL | 0.65cm | 4.5cm | 5cm |
Pack of 6 for $103.00
Case of 4 PK for $412.00 |
|
S34402
![]() |
Kimble
2895055/EMD |
36 mL | 0.8cm | 5.5cm | 6.3cm |
Pack of 12 for $122.00
Case of 4 PK for $488.00 |
|
S34405
![]() |
Kimble
2895090/EMD |
150 mL | 0.9cm | 9cm | 9.7cm |
Pack of 6 for $86.00
Case of 4 PK for $344.00 |
|
S34403
![]() |
Kimble
2895065/EMD |
60 mL | 0.8cm | 6.5cm | 6.3cm |
Pack of 12 for $248.00
Case of 4 PK for $992.00 |
|
S34404
![]() |
Kimble
2895075/EMD |
90 mL | 0.8cm | 7.5cm | 7.5cm |
Pack of 12 for $180.00
Case of 4 PK for $720.00 |
|
Specifications
58° | |
ASTM E1095, Type I, Class A | |
Borosilicate Glass | |
Provides faster, more effective filtering |