Kimberly-Clark™ Professional Basic™ Lab Coats Available on GSA/VA Contract for Federal Government customers only.

For light fluid contact

Manufacturer: Kimberly-Clark™ Professional 10124

 View more versions of this product

Catalog No. 19-043061

  Check Availability
Add to Cart

    Specifications

    Material SMS Fabric
    No. of Pockets 2.0
    Size 2X-Large
    For Use With (Application) Suited for light fluid contact
    Collar Type Traditional
    Size (English) Chest 54 to 60 in.
    Thickness 3-ply
    Closure Type Snap Front
    Color White
    Construction 3-ply SMS
    Cuff Style Knit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit