Learn More
Kimberly-Clark™ Professional Classic™ Surgical Masks
Manufacturer:  Kimberly-Clark™ Professional 49215
 View more versions of this product
Catalog No. 19-166-984
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Green
Tie closures, film strip, Derma-Touch™ tape
Regular
Pleated
0.1 (PFE), 3 (BFE)μ
3-ply construction
Fabric
≥96% (BFE), ≥97% (PFE)
No