Kimberly-Clark™ Professional Classic™ Surgical Masks
Manufacturer: Kimberly-Clark™ Professional 59928
 View more versions of this product
Catalog No. 19-166-988
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Filter Efficiency ≥96% (BFE), ≥97% (PFE)
    Color White
    Construction 3-ply construction
    Style Pleated
    Sterile No
    Pore Size 0.1 (PFE), 3 (BFE)μ
    Includes Tie closures
    Material Fabric
    Size Regular
  • Description & Specifications
Related Products
Product Content Feedback