Learn More
Kimberly-Clark Professional™ Comfort Latex Powder-Free Exam Gloves Get IT Item
Provides safety against hazardous substances and reduce risk of cross-contamination
Manufacturer:  Kimberly-Clark Professional™ 43434
 View more versions of this product
Catalog No. 19-169-064
Add to cart
Alert:
Caution: This product contains natural rubber latex, which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established
Description
Description
Provides safety against hazardous substances and reduce risk of cross-contamination
Certifications
510K approved for medical use
Compliance
Meets ASTM™ D3578 standards; FDA 510(k) approved for medical use.
Specifications
Specifications
Beaded/Rolled
Large
ASTM D3578, FDA 510(k)
Ambidextrous
5 mil