Kimberly-Clark™ Professional Coreless Double Roll Bath Tissue Dispenser Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Kimberly-Clark™ Professional 09604

 View more versions of this product

Catalog No. 19-813-850

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ADA Standard
    Color Smoke grey
    For Use With (Application) Provides built-in anti-microbial protection against most common stain and odor-causing bacteria
    Material Stainless Steel
    Product Type Coreless Double Roll Bathroom Tissue Dispenser

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit