Learn More
Kimberly-Clark Professional™ Filtermask Standard Face Mask Available on GSA/VA Contract for Federal Government customers only.
Kimberly-Clark™ Professional Filtermask Standard Face Mask is appropriate for situations where exposure to blood and/or bodily fluids is not a risk
Manufacturer:  Kimberly-Clark Professional™ 37525
Catalog No. 19-166-979
Add to cart
Description
Description
  • Protective three-layer construction
  • Pouch style with ties
  • Materials and donning attachments are sonically bonded
  • Mask is natural rubber latex-free
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Green
Pouch
3-ply
≥96% (BFE), ≥97% (PFE)
0.1 (PFE), 3 (BFE)μ
Ties