Learn More
Kimberly-Clark Professional™ Fluidshield™ Fog-Free Surgical Masks
Kimberly-Clark™ Professional Fluidshield™ Fog-Free Surgical Masks are designed to provide the maximum level of fluid resistance recognized by ASTM F2100-11
Supplier: Kimberly-Clark Professional™ 48207
This item is not returnable.
View return policy
Description
Kimberly-Clark™ Professional Fluidshield™ Fog-Free Surgical Masks are designed to provide the maximum level of fluid resistance recognized by ASTM F2100-11
- Ideal option if workers are exposed to heavy fluids
- Pleated
- Features surgical ties for a secure fit
- Made with a breathable film and So Soft lining
- Constructed with a combination of materials to resist fluid penetration upon impact
- Fog-free and made with a foam strip for extra comfort
Specifications
Medical/Procedural Face Mask | |
Orange | |
Pleated | |
48207 |
ASTM Level 3 | |
Ties | |
0.99 |