Kimberly-Clark™ Professional Fluidshield™ PCM2000 Facemasks
Manufacturer: Kimberly-Clark™ Professional 47707
 View more versions of this product
Catalog No. 18-999-4805
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Filter Efficiency 99%
    Color Orange
    Style Pouch
    Pore Size 0.1 (PFE), 3 (BFE)μ
    Includes Polyurethane headbands, earloops
    Size Regular
  • Description & Specifications
Related Products
Product Content Feedback