Kimberly-Clark™ Professional Fluidshield™ PCM2000 Facemasks
Manufacturer: Kimberly-Clark™ Professional 47700
 View more versions of this product
Catalog No. 19-169-161
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Filter Efficiency 99%
    Color Blue
    Style Pouch
    Includes Polyurethane headbands, earloops
    Size Regular
    Pore Size 0.1 (PFE), 3 (BFE)μ
  • Description & Specifications
Related Products
Product Content Feedback