Learn More
Kimberly-Clark Professional™ Impervious Gowns
Impervious gown made for comfort
Manufacturer:  Kimberly-Clark Professional™ 69601
 View more versions of this product
Catalog No. 19-065-278
Add to cart
Description
Description
  • Non-sterile
  • Film laminate
  • Exposure anticipated: potential for soaking for heavy fluid contact in front
  • Knit cuffs
  • Open back
Specifications
Specifications
Non-sterile
Knit
Film Laminate
Safety
Blue
2X-Large