Learn More
Kimberly-Clark Professional Impervious Gowns
Impervious gown made for comfort
Catalog Number | Mfr. No. | Color | Material | Size | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Material | Size | Sterility | Price | Quantity & Availability | ||
19-065-278
|
Kimberly-Clark Professional
69601 |
Blue | Film Laminate | 2X-Large | Non-sterile |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Non-sterile
- Film laminate
- Exposure anticipated: potential for soaking for heavy fluid contact in front
- Knit cuffs
- Open back
Specifications
Knit |