Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ Sanitouch™ Hard Roll Towel Dispenser

No-touch dispensing minimizes the spread of dirt and germs

Supplier:  Kimberly-Clark Professional™ 09990

Catalog No. 19-812-937

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Description

Description

  • Automatically advances and cuts a nominal 30.48cm (12 inch) of towel when pulling exposed towel
  • Includes emergency-feed knob and choice of key-activated spring lock or push button operation
  • Dispenses one 20.32cm (8 inch) diameter roll with 3.82 cm (1.5 inch) core
  • Made of high-impact plastic
  • Smoked transparent cover with a gray back

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Towel Dispenser
16.3 x 4 x 10.2 in. (41.4 x 27 x 25.9 cm)
41.4 cm
27 cm
25.9 cm
For improved hygiene
Emergency-feed knob and choice of key-activated spring lock or push button operation
Smoke Gray
16.3 in.
4 in.
10.2 in.
Plastic
Scott™ or Kleenex™ hard roll towels
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit