Learn More
Kimberly-Clark Professional™ Jackson Safety™ R10 Dual-Valve N95 Particulate Respirator
The innovative, adjustable comfort strap design is preferred over the headstraps of leading N95 respirators
Manufacturer:  Kimberly-Clark Professional™ 64260
Catalog No. 19-413-200
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Dual exhalation valves designed and positioned to help reduce eyewear fogging
  • Highly visible color-coded valve/strap attachment
  • Soft, comfortable foam nose pad with a cloth layer and bendable nose clip
  • Latex-free
  • Color-coded blue
Specifications
Specifications
Safety
Blue
N95 Filter
Dual valve
Bendable nose pad
Each
White, Gray
NIOSH
Grinding, sanding, sweeping, wood working
Yes
Particulate Respirator
Universal