Learn More
Kimberly-Clark™ Professional Jackson Safety™ BC100 Bump Caps
Kimberly-Clark™ Professional Jackson Safety™ BC100 Bump Caps offer protection from minor bumps and scalp lacerations
Supplier:  Kimberly-Clark™ Professional 14809
Encompass
 View more versions of this product
Catalog No. 19-150-1560
Only null left
Add to Cart
Description
Description
  • Lightweight for comfort
  • Made of molded HDPE (high-density polyethylene)
  • 4-point injection molded system includes an absorbent brow pad
  • Pinlock sizing system can accommodate head sizes 6.5 to 8.25
  • Can be made compatible with a face shield for additional coverage
  • Replacement sweat bands and suspension inserts available
Specifications
Specifications
Yellow
Low-profile Bump Cap
Adjustable (Pin-lock sizing)
HDPE
12/Pk.
4-Point Injection Molded
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.