Learn More
Kimberly-Clark Professional™ KleenGuard™ G40 Smooth Nitrile Coated Multi-Purpose Gloves
General purpose gloves designed for conditions which require protection, strength, grip and dexterity in industrial environments. Nitrile coating offers a secure grip while the seamless knit nylon back offers a comfortable fit.
Specifications
Coating Material Nitrile
Material Nylon
Exterior Finish No Texture
Color Black
Glove Cuff Style Knit
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-005-420
View Documents
Kimberly-Clark Professional™
38428
Small
Dozen of 12 for $124.50
Case of 5 DZ for $592.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-005-421
View Documents
Kimberly-Clark Professional™
38429
Medium
Dozen of 12 for $124.50
Case of 5 DZ for $592.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-005-422
View Documents
Kimberly-Clark Professional™
38430
Large
Dozen of 12 for $124.50
Case of 5 DZ for $592.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-005-423
View Documents
Kimberly-Clark Professional™
38431
X-Large
Dozen of 12 for $124.50
Case of 5 DZ for $592.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-005-424
View Documents
Kimberly-Clark Professional™
38432
2X-Large
Dozen of 12 for $124.50
Case of 5 DZ for $592.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Designed for conditions which require protection, strength, grip and dexterity in industrial environments
  • Nitrile coating enhances grip capabilities and protects against liquid permeation
  • Seamless knit nylon back is comfortable and breathable
  • Dark color helps to conceal dirt for extended wear
Specifications
Specifications
Nitrile
No Texture
Knit
Nylon
Black
Kleengaurd™ G40