Kimberly-Clark™ Professional Jackson Saftey™ G60 Level 2 Polyurethane Coated Cut Resistant Gloves

Great choice for working environments that pose a risk of minor cuts and abrasions

Manufacturer: Kimberly-Clark™ Professional 38639

 View more versions of this product

Catalog No. 19-169-404

  N/A
Add to Cart

Alert

This product contains natural rubber latex, which may cause allergic reactions. This product should not be worn by, or exposed to, individuals allergic to natural rubber latex. Safe use of this product on or by latex-sensitive individuals has no t been established.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Hem Color Brown
    For Use With (Application) Metal fabrication, Glass handling, automotive assembly, stamping and assembly
    Cut Resistance Rating Level 2
    Length (English) 10.2 in.
    Length (Metric) 25.9cm
    Material Kevlar™
    Size 2X-Large
    Coating Material Polyurethane
    Color Yellow, gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit