Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ G60 Knuckle Coated High-Visibility Cut-Resistant Gloves
Click to view available options
Length (English):
8.4 in.
8.9 in.
9.3 in.
9.6 in.
10 in.
Length (Metric):
21.3 cm
22.6 cm
23.6 cm
24.3 cm
25.4 cm
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- EN 388 Level 3 / ANSI Level 2 Cut Resistance
- EN 388 Level 4 abrasion and Level 4 tear resistance
- Certified to meet the EN 420 standards
- Bright orange fiber aids in overall safety compliance and awareness of how close a hand is in proximity to potential dangers
- Made of ultra-high molecular weight polyethylene (UHMWPE) yarn with nylon and stretch fiber, which provides both cut resistance and a comfortable fit
- Polyurethane coating enhances dry and oil grip and hides dirt for longer wear
- Dark coating helps hide dirt for longer wear
- Each pair has a dedicated right and left glove for a snug fit
- Knuckle coating helps keep hands and gloves protected from liquids and oils to enforce compliance and improve glove longevity
Specifications
ANSI Abrasion Rating | ANSI 4 |
Certifications/Compliance (Europe) | EN 420 |
Color | Black, Orange |
Cut Resistance Rating | ANSI A3 |
Material | Dyneema™, Nylon, Lycra™ |
Product Type | Cut Resistant Gloves |
Size | Large |
EN388 Standard | EN 388 |
For Use With (Application) | Metal fabrication, Glass handling, automotive assembly, stamping and assembly |
Glove Cuff Style | Knit |
Show More |